Thermo Scientific Chemicals

Sodium tetrafluoroborate, 95%, Thermo Scientific Chemicals

Catalog number: 011562.36
500 g, Each
Thermo Scientific Chemicals

Sodium tetrafluoroborate, 95%, Thermo Scientific Chemicals

Catalog number: 011562.36
500 g, Each
Quantity
Catalog number: 011562.36
also known as 011562-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13755-29-8
IUPAC Name
sodium tetrafluoroboranuide
Molecular Formula
BF4Na
InChI Key
KGJZTOFHXCFQIV-UHFFFAOYSA-N
SMILES
[Na+].F[B-](F)(F)F
Assay (unspecified)
94.1% min

Description

Fluorinating agentSodium tetrafluoroborate is used in fluxes, metal finishing and in plating circuits. It is used as a starting material in the preparation of ionic liquids as well as boron trifluoride. It acts as an efficient catalyst for the synthesis of 3,4-dihydropyrimidin-2(1H)-ones and thiones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text