Magnesium nitrate hexahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals
Magnesium nitrate hexahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Magnesium nitrate hexahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

As fertilizer, in manufacture of ammonium nitrate
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
500 g
2 kg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 011564.18
Price (USD)/ Each
18.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
22.30 
Save 3.65 (16%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
18.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
22.30 
Save 3.65 (16%)
Add to cart
Magnesium nitrate hexahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals
Catalog number011564.18
Price (USD)/ Each
18.65
Special Offer
Online exclusive
Ends: 31-Dec-2024
22.30 
Save 3.65 (16%)
-
Add to cart
Chemical Identifiers
CAS13446-18-9
IUPAC Namemagnesium(2+) hexahydrate dinitrate
Molecular FormulaH12MgN2O12
InChI KeyMFUVDXOKPBAHMC-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.[Mg++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentFe 5ppm
Impurity contentK 0.005%
Impurity contentMn 5ppm
Impurity contentNH₄ 0.003%
Impurity contentPO₄ 5ppm
View more

It is used in printing, chemical, agriculture and ceramics industries. It is also used in the detection of selenium in plasma samples, and concentration of nitric acid. Its fertilizer grade has 10.5% nitrogen and 9.4% magnesium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only