Chromium(III) nitrate nonahydrate, 98.5%, Thermo Scientific Chemicals
Chromium(III) nitrate nonahydrate, 98.5%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Chromium(III) nitrate nonahydrate, 98.5%, Thermo Scientific Chemicals

For Use As a Catalyst, Corrosion Inhibitor, in Textile Printing
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
25 g
250 g
Catalog number 011565.A1
also known as 011565-A1
Price (USD)/ Each
164.65
Online exclusive
183.00 
Save 18.35 (10%)
-
Add to cart
Quantity:
1 kg
Request bulk or custom format
Price (USD)/ Each
164.65
Online exclusive
183.00 
Save 18.35 (10%)
Add to cart
Chromium(III) nitrate nonahydrate, 98.5%, Thermo Scientific Chemicals
Catalog number011565.A1
Price (USD)/ Each
164.65
Online exclusive
183.00 
Save 18.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7789-02-8
IUPAC Namechromium(3+) trinitrate
Molecular FormulaCrN3O9
InChI KeyPHFQLYPOURZARY-UHFFFAOYSA-N
SMILES[Cr+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.5% min
Chromium nitrate is used in the manufacture of alkali metal-free catalysts and in preservation. It is used in the preparation of chrome catalysts, in textile printing operations, and as a corrosion inhibitor. Chromium(III) nitrate as a source of chromium is added to food supplements for nutritional purposes .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only