Thermo Scientific Chemicals

Ammonium fluoride, ACS, 98.0% min, Thermo Scientific Chemicals

Catalog number: 011568.30
250 g, Each
Thermo Scientific Chemicals

Ammonium fluoride, ACS, 98.0% min, Thermo Scientific Chemicals

Catalog number: 011568.30
250 g, Each
Quantity
Catalog number: 011568.30
also known as 011568-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12125-01-8
IUPAC Name
ammonium hydrogen difluoride
Molecular Formula
F2H5N
InChI Key
KVBCYCWRDBDGBG-UHFFFAOYSA-N
SMILES
[H+].[NH4+].[F-].[F-]
Assay (unspecified)
98.0% NH₄F min
Impurity content
Cl 0.001%
Impurity content
Fe 5ppm
Insoluble material
0.005%
Impurity content
SO₄ 0.005%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text