Thermo Scientific Chemicals

Tin(IV) chloride hydrate, 98%, Thermo Scientific Chemicals

Catalog number: 011570.22
100 g, Each
Thermo Scientific Chemicals

Tin(IV) chloride hydrate, 98%, Thermo Scientific Chemicals

Catalog number: 011570.22
100 g, Each
Quantity
Catalog number: 011570.22
also known as 011570-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10026-06-9
IUPAC Name
tin(4+) pentahydrate tetrachloride
Molecular Formula
Cl4H10O5Sn
InChI Key
KHMOASUYFVRATF-UHFFFAOYSA-J
SMILES
O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Sn+4]
Assay (unspecified)
97.6% min

Description

Tin(IV) chloride hydrate is used in the formation of a xerogel via a cyanogel, a gel that has potential use in solid-state gas sensors. It is involved in the preparation of Indium tin oxide thin films by chemical vapor deposition method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text