Rhodium wire, 0.5mm (0.02in) dia, 99.8% (metals basis), Thermo Scientific Chemicals
Rhodium wire, 0.5mm (0.02in) dia, 99.8% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Rhodium wire, 0.5mm (0.02in) dia, 99.8% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 cm
7.5 cm
10 cm
30 cm
Catalog number 011576.BW
also known as 011576-BW
Price (USD)/ Each
-
Quantity:
50 cm
Request bulk or custom format
Chemical Identifiers
CAS7440-16-6
IUPAC Namerhodium
Molecular FormulaRh
InChI KeyMHOVAHRLVXNVSD-UHFFFAOYSA-N
SMILES[Rh]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities≤2000ppm
Assay (unspecified)≥99.8% (metals basis)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only