Thermo Scientific Chemicals

Antimony(III) oxide, 99.6% min, Thermo Scientific Chemicals

Catalog number: 011579.18
50 g, Each
Thermo Scientific Chemicals

Antimony(III) oxide, 99.6% min, Thermo Scientific Chemicals

Catalog number: 011579.18
50 g, Each
Quantity
Catalog number: 011579.18
also known as 011579-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1309-64-4
IUPAC Name
diantimony(3+) trioxidandiide
Molecular Formula
O3Sb2
InChI Key
GHPGOEFPKIHBNM-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Sb+3].[Sb+3]
Assay (unspecified)
99.6% min

Description

It finds applications in flame retardants, catalysts, batteries, and alloys. It acts as an opacifying agent in glasses, ceramics and enamels. It is used as a catalyst in the manufacture of PET plastic (polyethylene terephthalate), and the vulcanization of rubber.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text