Thermo Scientific Chemicals

Sodium dihydrogen phosphate monohydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Catalog number: 011591.A1
1 kg, Each
Thermo Scientific Chemicals

Sodium dihydrogen phosphate monohydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Catalog number: 011591.A1
1 kg, Each
Quantity
Catalog number: 011591.A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10049-21-5
IUPAC Name
sodium phosphoric acid hydrate
Molecular Formula
H5NaO5P
InChI Key
BBMHARZCALWXSL-UHFFFAOYSA-N
SMILES
O.[Na+].OP(O)(O)=O
Heavy Metals
(ICP-OES): ≤0.001%
Impurity content
Ca: ≤0.005%
Impurity content
Cl: ≤5ppm
Impurity content
Fe: ≤0.001%
Impurity content
K: ≤0.01%

Description

Sodium phosphate monobasic monohydrate is used as sequestrant, emulsifier, mordant in dyeing, reagent and buffer in foods and analytical chemistry. It is applied as a fireproofing agent and for weighting silk in tanning. It is employed in manufacturing of enamels, ceramics, detergents, boiler compounds, in soldering and brazing instead of borax.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text