Potassium hydrogen phosphate, ACS, 98.0% min, Thermo Scientific Chemicals
Potassium hydrogen phosphate, ACS, 98.0% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Potassium hydrogen phosphate, ACS, 98.0% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
250 g
1 kg
Catalog number 011593.A7
also known as 011593-A7
Price (USD)/ Each
494.65
Online exclusive
549.00 
Save 54.35 (10%)
-
Add to cart
Quantity:
5 kg
Request bulk or custom format
Price (USD)/ Each
494.65
Online exclusive
549.00 
Save 54.35 (10%)
Add to cart
Potassium hydrogen phosphate, ACS, 98.0% min, Thermo Scientific Chemicals
Catalog number011593.A7
Price (USD)/ Each
494.65
Online exclusive
549.00 
Save 54.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7758-11-4
IUPAC Namedipotassium hydrogen phosphate
Molecular FormulaHK2O4P
InChI KeyZPWVASYFFYYZEW-UHFFFAOYSA-L
SMILES[K+].[K+].OP([O-])([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity contentCl 0.003%
Impurity contentNitrogen compounds (as N) 0.001%
Impurity contentSO₄ 0.005%
Assay (unspecified)98.0% K₂HPO₄ min
Insoluble material0.01%
View more
It is used as a fertilizer, food additive, buffering agent, in molecular biology, biochemistry and chromatography. It is useful to increase the partition coefficients of amino acids in aqueous two-phase systems. It plays an important role for increase in rice grain yield by spraying chemicals including ferric chloride, dipotassium hydrogen phosphate and salicylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only