Thermo Scientific Chemicals

Potassium chloride, ACS, 99.0-100.5%, Thermo Scientific Chemicals

Catalog number: 011595.30
250 g, Each
Thermo Scientific Chemicals

Potassium chloride, ACS, 99.0-100.5%, Thermo Scientific Chemicals

Catalog number: 011595.30
250 g, Each
Quantity
Catalog number: 011595.30
also known as 011595-30
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7447-40-7
IUPAC Name
potassium chloride
Molecular Formula
ClK
InChI Key
WCUXLLCKKVVCTQ-UHFFFAOYSA-M
SMILES
[Cl-].[K+]
Impurity content
(Barium, Ba) : Passes test (limit about 0.001%)
Impurity content
(Iodide, I) : 0.002% max
Impurity content
(Iron, Fe) : 3ppm max
Impurity content
(Magnesium, Mg) : 0.001% max
Impurity content
(Phosphate, PO₄) : 5ppm max

Description

Potassium chloride is used to replenish electrolytes and restore water electrolyte balance in treating hypokalemia. It is also used in buffers, fertilizers and explosives and finds useful in medicine, scientific applications, and food processing. It plays an important role for the extraction and solubilization of proteins. It acts as a fire extinguishing agent in portable and wheeled fire extinguishers. It may be used as a softener unit in household water, which is an alternative to sodium chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • In buffer solution, electrode cells, photography, lab reagent, fertilizer, spectroscopy
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text