Thermo Scientific Chemicals

Magnesium sulfate heptahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Catalog number: 011596.22
100 g, Each
Thermo Scientific Chemicals

Magnesium sulfate heptahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Catalog number: 011596.22
100 g, Each
Quantity
Catalog number: 011596.22
also known as 011596-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10034-99-8
IUPAC Name
magnesium(2+) heptahydrate sulfate
Molecular Formula
H14MgO11S
InChI Key
WRUGWIBCXHJTDG-UHFFFAOYSA-L
SMILES
O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O
Assay (unspecified)
98.0-102.0% MgSO₄?7H₂O
Impurity content
Mn 5ppm
Impurity content
Na 0.005%
Impurity content
NH₄ 0.002%
Impurity content
NO₃ 0.002%

Description

Magnesium sulfate produced by appropriate process finds use in several pharmaceutical preparations of magnesium, usually known as epsom salt. It is used as a micronutrient in agriculture, brewing salt in production of certain alcoholic products, electrolyte to prepare metal salts, coagulant, and ingredient in bath salts for use in floatation therapy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text