Thermo Scientific Chemicals

Ammonium hydrogen phosphate, ACS, 98.0% min, Thermo Scientific Chemicals

Catalog number: 011597.A1
1 kg, Each
Thermo Scientific Chemicals

Ammonium hydrogen phosphate, ACS, 98.0% min, Thermo Scientific Chemicals

Catalog number: 011597.A1
1 kg, Each
Quantity
Catalog number: 011597.A1
also known as 011597-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7783-28-0
IUPAC Name
phosphoric acid diamine
Molecular Formula
H9N2O4P
InChI Key
MNNHAPBLZZVQHP-UHFFFAOYSA-N
SMILES
N.N.OP(O)(O)=O
Assay (unspecified)
98.0% (NH₄)₂HPO₄ min
Impurity content
Ca 0.001%
Impurity content
Cl 0.001%
Impurity content
Fe 0.001%
Impurity content
K 0.005%

Description

Ammonium hydrogen phosphate is commonly used as a fertilizer. It can be used as fire retardant, in fireproofing textiles, paper and wood. It finds applications in vegetable fibers, impregnating lamp wicks, soldering metals, purifying sugar, in yeast cultures and corrosion inhibitors. It is used in the treatment of carbonate stones used in architectural, cultural heritage and optical materials. It is also used as a flux for soldering copper, tin and zinc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text