Ammonium dihydrogen phosphate, ACS, 98.0% min, Thermo Scientific Chemicals
Ammonium dihydrogen phosphate, ACS, 98.0% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium dihydrogen phosphate, ACS, 98.0% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
500 g
Catalog number 011598.A3
also known as 011598-A3
Price (USD)/ Each
178.00
-
Add to cart
Quantity:
2 kg
Request bulk or custom format
Price (USD)/ Each
178.00
Add to cart
Ammonium dihydrogen phosphate, ACS, 98.0% min, Thermo Scientific Chemicals
Catalog number011598.A3
Price (USD)/ Each
178.00
-
Add to cart
Chemical Identifiers
CAS7722-76-1
IUPAC Namephosphoric acid amine
Molecular FormulaH6NO4P
InChI KeyLFVGISIMTYGQHF-UHFFFAOYSA-N
SMILESN.OP(O)(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity content≤0.005% [Potassium (K)]
Impurity content≤0.005% [Sodium (Na)]
Impurity content≤0.01% [Sulfate (SO4)]
Insoluble material≤0.005%
Heavy Metals (as Pb)≤5 ppm
View more
Preparation of baking powder with sodium bicarbonate, in fermentation.Ammonium dihydrogen phosphate is widely utilized as a fertilizer, which provides phosphorous and nitrogen for plants. It is used as a dry fire extinguisher. It is a well known nonlinear optical material for various optoelectronic applications. It is also used in an emerald green, amethyst and aquamarine crystal growing box kit for children.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only