Thermo Scientific Chemicals

Copper(I) bromide, 98%, Thermo Scientific Chemicals

Catalog number: 011605.A1
1 kg, Each
Thermo Scientific Chemicals

Copper(I) bromide, 98%, Thermo Scientific Chemicals

Catalog number: 011605.A1
1 kg, Each
Quantity
Catalog number: 011605.A1
also known as 011605-A1
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7787-70-4
IUPAC Name
λ¹-copper(1+) bromide
Molecular Formula
BrCu
InChI Key
NKNDPYCGAZPOFS-UHFFFAOYSA-M
SMILES
[Cu+].[Br-]
Appearance (Color)
Green
Form
Powder
Assay (special)
≥ 97.5%

Description

It finds many applications in organic synthesis, as a catalyst and a reagent. It is used as a catalyst in Sandmeyer reaction for preparing aryl bromides from arylamines through diazonium salts. It forms complexes with Lewis bases, such as dimethylsulfide, which is a precursor to organocopper reagents. Reactions involving Atom Transfer Radical Polymerization (ATRP) and Cu-catalyzed Cross-Dehydrogenative Couplings (CDC) are catalyzed by CuBr complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text