Silver(II) fluoride, 98+%, Thermo Scientific Chemicals
Silver(II) fluoride, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Silver(II) fluoride, 98+%, Thermo Scientific Chemicals

Moisture and Light Sensitive
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 g
10 g
50 g
Catalog number 011610.04
also known as 011610-04
Price (USD)/ Each
41.65
Online exclusive
45.60 
Save 3.95 (9%)
-
Add to cart
Quantity:
2 g
Request bulk or custom format
Price (USD)/ Each
41.65
Online exclusive
45.60 
Save 3.95 (9%)
Add to cart
Silver(II) fluoride, 98+%, Thermo Scientific Chemicals
Catalog number011610.04
Price (USD)/ Each
41.65
Online exclusive
45.60 
Save 3.95 (9%)
-
Add to cart
Chemical Identifiers
CAS7783-95-1
IUPAC Namesilver(II) fluoride
Molecular FormulaAgF2H2
InChI KeyDOVBQBITXCIRPH-UHFFFAOYSA-N
SMILESF.F.[Ag]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)98.1% min
Appearance (Color)Gray to brown
FormCrystalline
Silver(II) fluoride is used as a fluorinating and oxidizing agent. It is also used in the preparation of organic perfluorocompounds and in the fluorination of aromatic compounds. In addition, it is also used in pharmacy and electroplating industry. It serves as an intermediate in the catalysis of gaseous reactions with fluorine by silver. It is utilized in the oxidation of xenon to xenon difluoride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only