Zinc perchlorate hexahydrate, Reagent Grade, Thermo Scientific Chemicals
Zinc perchlorate hexahydrate, Reagent Grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc perchlorate hexahydrate, Reagent Grade, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Catalog number 011614.22
also known as 011614-22
Price (USD)/ Each
28.65
Online exclusive
32.00 
Save 3.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
28.65
Online exclusive
32.00 
Save 3.35 (10%)
Add to cart
Zinc perchlorate hexahydrate, Reagent Grade, Thermo Scientific Chemicals
Catalog number011614.22
Price (USD)/ Each
28.65
Online exclusive
32.00 
Save 3.35 (10%)
-
Add to cart
Chemical Identifiers
CAS10025-64-6
IUPAC Namezinc(2+) hexahydrate diperchlorate
Molecular FormulaCl2H12O14Zn
InChI KeyPADPILQDYPIHQQ-UHFFFAOYSA-L
SMILESO.O.O.O.O.O.[Zn++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Zn 16.7-18.0%

Zinc perchlorate hexahydrate, efficiently catalyses the esterification between nearly equimolar amounts of carboxylic acids and alcoholsZinc perchlorate hexahydrate [Zn (ClO4) 2· 6H2O] was found to be a highly efficient catalyst for acylation of electron deficient phenols, sterically hindered alcohols and electron deficient and sterically hindered amines. Zinc (II) perchlorate hexahydrate has been found to be a new and efficient catalyst for conjugate addition of thiols to α, β-unsaturated ketones under solvent-free conditions at room temperature.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only