Sodium perchlorate is used in chemical analysis and in explosive materials, fireworks, and rocket motors. It is the precursor for the preparation of other perchlorate salts. It is also employed as a chaotrophic agent in DNA extraction and hybridization reactions in molecular biology. It prevents the binding of phenolic compounds to RNA during extraction. In addition, it is used in batteries, plastics and in rubber products. It is also used as a nonreactive electrolyte. Further, it is an eluent component used for analysis of biological samples by liquid chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.