Thermo Scientific Chemicals

Sodium perchlorate, anhydrous, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Catalog number: 011623.36
500 g, Each
Thermo Scientific Chemicals

Sodium perchlorate, anhydrous, ACS, 98.0-102.0%, Thermo Scientific Chemicals

Catalog number: 011623.36
500 g, Each
Quantity
Informational:Informational:This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number: 011623.36
also known as 011623-36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7601-89-0
IUPAC Name
sodium perchlorate
Molecular Formula
ClNaO4
InChI Key
BAZAXWOYCMUHIX-UHFFFAOYSA-M
SMILES
[Na+].[O-][Cl](=O)(=O)=O
Assay (unspecified)
98.0-102.0% NaClO₄
Heavy Metals
(ICP-OES) 5ppm
Impurity content
Ca 0.02%
Insoluble material
0.005%
Impurity content
Cl 0.003%

Description

Sodium perchlorate is used in chemical analysis and in explosive materials, fireworks, and rocket motors. It is the precursor for the preparation of other perchlorate salts. It is also employed as a chaotrophic agent in DNA extraction and hybridization reactions in molecular biology. It prevents the binding of phenolic compounds to RNA during extraction. In addition, it is used in batteries, plastics and in rubber products. It is also used as a nonreactive electrolyte. Further, it is an eluent component used for analysis of biological samples by liquid chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text