Thermo Scientific Chemicals

Silver perchlorate monohydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 011624.09
10 g, Each
Thermo Scientific Chemicals

Silver perchlorate monohydrate, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 011624.09
10 g, Each
Quantity
Catalog number: 011624.09
also known as 011624-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14242-05-8
IUPAC Name
silver(1+) perchlorate
Molecular Formula
AgClO4
InChI Key
YDHABVNRCBNRNZ-UHFFFAOYSA-M
SMILES
[Ag+].[O-][Cl](=O)(=O)=O
Total Metal Impurities
0.1% max.

Description

Silver perchlorate can be used in the manufacturing of explosives. It is generally used as a source of silver ions during silver nanoparticles synthesis. It is used as a catalyst in organic chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text