Thermo Scientific Chemicals

Nickel(II) perchlorate hexahydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 011631.14
25 g, Each
Thermo Scientific Chemicals

Nickel(II) perchlorate hexahydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 011631.14
25 g, Each
Quantity
Catalog number: 011631.14
also known as 011631-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
13520-61-1
IUPAC Name
nickel(2+) hexakis(methane) diperchlorate
Molecular Formula
C6H24Cl2NiO8
InChI Key
GWPPTBKPEOUCMN-UHFFFAOYSA-L
SMILES
C.C.C.C.C.C.[Ni++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Assay (unspecified)
Ni 15.3û17.0%

Description

Nickel(II) perchlorate hexahydrate is used as a starting material for homometallic, trinuclear heteroscorpionate complexes applied in studies of electronic and magnetic properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text