Thermo Scientific Chemicals

Lead(II) perchlorate trihydrate, ACS, 97.0-102.0%, Thermo Scientific Chemicals

Catalog number: 011640.22
100 g, Each
Thermo Scientific Chemicals

Lead(II) perchlorate trihydrate, ACS, 97.0-102.0%, Thermo Scientific Chemicals

Catalog number: 011640.22
100 g, Each
Quantity
Catalog number: 011640.22
also known as 011640-22
Price (USD)
Have Questions?

Chemical Identifiers

CAS
13453-62-8
IUPAC Name
λ²-lead(2+) trihydrate diperchlorate
Molecular Formula
Cl2H6O11Pb
InChI Key
KKGGEAQRICYXNM-UHFFFAOYSA-L
SMILES
O.O.O.[Pb++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Impurity content
Fe: ≤0.001%
Appearance (Color)
White to cream
Form
Powder or crystals or crystalline powder
Assay (unspecified)
97.0-102.0%
Insoluble material
≤0.005%

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text