Thermo Scientific Chemicals

Lanthanum(III) perchlorate hexahydrate, Thermo Scientific Chemicals

Catalog number: 011641.14
25 g, Each
Thermo Scientific Chemicals

Lanthanum(III) perchlorate hexahydrate, Thermo Scientific Chemicals

Catalog number: 011641.14
25 g, Each
Quantity
Catalog number: 011641.14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
14017-46-0
IUPAC Name
lanthanum(3+) hexahydrate triperchlorate
Molecular Formula
Cl3H12LaO18
InChI Key
DCEUOMQVLPJQAI-UHFFFAOYSA-K
SMILES
O.O.O.O.O.O.[La+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Assay (unspecified)
La 24.2-26.5%

Description

Lanthanum(III) perchlorate hexahydrate participates in the preparation of the Schiff base complexes of lanthanide (III) ions. When hydrated lanthanum perchlorate in ethanolic solution is treated with an ethanolic solution of terpyridine, [La(terpy)3](CIO4)3 is deposited as very small crystals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text