Thermo Scientific Chemicals

Iron(III) perchlorate hydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 011647.22
100 g, Each
Thermo Scientific Chemicals

Iron(III) perchlorate hydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 011647.22
100 g, Each
Quantity
Catalog number: 011647.22
also known as 011647-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
15201-61-3
IUPAC Name
iron(3+) triperchlorate
Molecular Formula
Cl3FeO12
InChI Key
LHOWRPZTCLUDOI-UHFFFAOYSA-K
SMILES
[Fe+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Assay (unspecified)
65.0 -80.0% Fe(ClO₄)₃

Description

Iron(III) perchlorate hydrate is used as a starting material for the synthesis of a tris-salicylate Fe(III) complex having bidentate chelating units that allow two coordination modes. It is also used as a catalyst for synthesis of fullerene-fused lactones, α-Carbonyl furans via one-pot cyclization, fullerodioxolanes via heterocyclization, dialylated indoles via double alkylation, aryl esters via oxidative esterification and as a chemical actinometer based on photolysis of ferrioxalate in presence of polyoxometalate in aqueous solution, as an oxidant for conducting polymer nanoparticles synthesized in ionic liquid by chemical polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text