Calcium perchlorate hydrate, Reagent Grade, Thermo Scientific Chemicals
Calcium perchlorate hydrate, Reagent Grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium perchlorate hydrate, Reagent Grade, Thermo Scientific Chemicals

Have Questions?
Quantity:
250 g
Catalog number 011655.30
also known as 011655-30
Price (USD)/ Each
57.65
Online exclusive
63.50 
Save 5.85 (9%)
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
57.65
Online exclusive
63.50 
Save 5.85 (9%)
Add to cart
Calcium perchlorate hydrate, Reagent Grade, Thermo Scientific Chemicals
Catalog number011655.30
Price (USD)/ Each
57.65
Online exclusive
63.50 
Save 5.85 (9%)
-
Add to cart
Chemical Identifiers
CAS13477-36-6
IUPAC Namecalcium diperchlorate
Molecular FormulaCaCl2O8
InChI KeyZQAOXERLHGRFIC-UHFFFAOYSA-L
SMILES[Ca++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Ca 12.2-13.5%
Calcium perchlorate hydrate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only