Thermo Scientific Chemicals

Ammonium perchlorate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 011658.30
250 g, Each
Thermo Scientific Chemicals

Ammonium perchlorate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 011658.30
250 g, Each
Quantity
Catalog number: 011658.30
Price (USD)
Have Questions?

Chemical Identifiers

CAS
7790-98-9
Insoluble material
0.005% max
Impurity content
Fe 0.0005% max
Impurity content
Chloride + Chlorate 0.003% max
Impurity content
SO₄ 0.001% max
pH
4.0-6.0

Description

Ammonium perchlorate is used to make rocket propellants, explosives, pyrotechnics, as an etching and engraving agent, and in analytical chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text