Thermo Scientific Chemicals

Aluminum perchlorate nonahydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 011659.22
100 g, Each
Thermo Scientific Chemicals

Aluminum perchlorate nonahydrate, Reagent Grade, Thermo Scientific Chemicals

Catalog number: 011659.22
100 g, Each
Quantity
Catalog number: 011659.22
also known as 011659-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
81029-06-3
IUPAC Name
aluminium(3+) nonahydrate triperchlorate
Molecular Formula
AlCl3H18O21
InChI Key
CYRDFMGMNQJULY-UHFFFAOYSA-K
SMILES
O.O.O.O.O.O.O.O.O.[Al+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
Impurity content
0.1% max

Description

Aluminum perchlorate nonahydrate is used as an intermediate in both pharmaceutical industry and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text