Aluminum perchlorate nonahydrate, Reagent Grade, Thermo Scientific Chemicals
Aluminum perchlorate nonahydrate, Reagent Grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum perchlorate nonahydrate, Reagent Grade, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 011659.36
also known as 011659-36
Price (USD)/ Each
150.65
Online exclusive
167.00 
Save 16.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
150.65
Online exclusive
167.00 
Save 16.35 (10%)
Add to cart
Aluminum perchlorate nonahydrate, Reagent Grade, Thermo Scientific Chemicals
Catalog number011659.36
Price (USD)/ Each
150.65
Online exclusive
167.00 
Save 16.35 (10%)
-
Add to cart
Chemical Identifiers
CAS81029-06-3
IUPAC Namealuminium(3+) nonahydrate triperchlorate
Molecular FormulaAlCl3H18O21
InChI KeyCYRDFMGMNQJULY-UHFFFAOYSA-K
SMILESO.O.O.O.O.O.O.O.O.[Al+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Impurity content0.1% max
Aluminum perchlorate nonahydrate is used as an intermediate in both pharmaceutical industry and chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only