Thermo Scientific Chemicals

Silver acetate, anhydrous, 99%, Thermo Scientific Chemicals

Catalog number: 011660.22
100 g, Each
Thermo Scientific Chemicals

Silver acetate, anhydrous, 99%, Thermo Scientific Chemicals

Catalog number: 011660.22
100 g, Each
Quantity
Catalog number: 011660.22
also known as 011660-22
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
563-63-3
IUPAC Name
silver(1+) acetate
Molecular Formula
C2H3AgO2
InChI Key
CQLFBEKRDQMJLZ-UHFFFAOYSA-M
SMILES
[Ag+].CC([O-])=O
Appearance (Color)
White to cream
Form
Microcrystalline powder or powder
Assay from Supplier's CofA
≥98.5% to ≤101.5% [UK sourced material]
Assay from Supplier's CofA
64.5 ± 0.5% Ag w/w [US sourced material]
Comment
Material sourced either in UK or USA

Description

It is a reagent in the laboratory as a source of silver ions lacking an oxidizing anion. It is a reagent for direct ortho-arylation, and for conversion of organohalogen compounds into alcohols. Woodward cis-hydroxylation reaction employs silver acetate and iodine for selective conversin of alkenes into cis-diols. Silver acetate is the more preferred reagent for facile carbonylation of primary and secondary amines. It is also employed in the preparation of highly reflective, conductive silvered polymer films.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text