Zinc iodide, 98%, Thermo Scientific Chemicals
Zinc iodide, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Zinc iodide, 98%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Catalog number 011661.18
also known as 011661-18
Price (USD)/ Each
71.65
Online exclusive
79.70 
Save 8.05 (10%)
-
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (USD)/ Each
71.65
Online exclusive
79.70 
Save 8.05 (10%)
Add to cart
Zinc iodide, 98%, Thermo Scientific Chemicals
Catalog number011661.18
Price (USD)/ Each
71.65
Online exclusive
79.70 
Save 8.05 (10%)
-
Add to cart
Chemical Identifiers
CAS10139-47-6
IUPAC Namezinc(2+) diiodide
Molecular FormulaI2Zn
InChI KeyUAYWVJHJZHQCIE-UHFFFAOYSA-L
SMILES[Zn++].[I-].[I-]
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)97.6% min
Zinc iodide is often used as an x-ray opaque penetrant in industrial radiography to improve the contrast between the damage and intact composite.n conjunction with osmium tetroxide ZnI2 is used as a stain in electron microscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only