Sodium hexanitritocobaltate(III), ACS, Thermo Scientific Chemicals
Sodium hexanitritocobaltate(III), ACS, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium hexanitritocobaltate(III), ACS, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number 011667.36
also known as 011667-36
Price (USD)/ Each
454.00
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
454.00
Add to cart
Sodium hexanitritocobaltate(III), ACS, Thermo Scientific Chemicals
Catalog number011667.36
Price (USD)/ Each
454.00
-
Add to cart
Chemical Identifiers
CAS14649-73-1
IUPAC Namecobalt(3+) trisodium hexakis(dioxoamine)
Molecular FormulaCoH6N6Na3O12
InChI KeyQLNJFTGEDYRKNP-UHFFFAOYSA-N
SMILES[Na+].[Na+].[Na+].[Co+3].[O-][NH+]=O.[O-][NH+]=O.[O-][NH+]=O.[O-][NH+]=O.[O-][NH+]=O.[O-][NH+]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Insoluble material0.02%
CommentSuitability for determination of potassium P.T.
Sodium hexanitritocobaltate(III) is used as primary and secondary intermediate in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only