Thermo Scientific Chemicals

Ammonium iodide, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 011668.36
500 g, Each
Thermo Scientific Chemicals

Ammonium iodide, ACS, 99.0% min, Thermo Scientific Chemicals

Catalog number: 011668.36
500 g, Each
Quantity
Catalog number: 011668.36
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12027-06-4
IUPAC Name
ammonium iodide
Molecular Formula
H4IN
InChI Key
UKFWSNCTAHXBQN-UHFFFAOYSA-N
SMILES
[NH4+].[I-]
Assay (unspecified)
99.0% NH₄I min
Residue on ignition
0.05%
Impurity content
Chloride and bromide (as Cl) 0.005%
Impurity content
PO₄ 0.001%
Impurity content
SO₄ 0.05%

Description

Appropriately manufactured ammonium iodide is used in pharmaceutical industries. It is used in photographic chemicals, some medications and several chemical industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text