Antimony pieces, 5mm (0.2in) & down, 99.9999% (metals basis), Thermo Scientific Chemicals
Antimony pieces, 5mm (0.2in) & down, 99.9999% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Antimony pieces, 5mm (0.2in) & down, 99.9999% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Catalog number 011676.09
also known as 011676-09
Price (USD)/ Each
157.00
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
157.00
Add to cart
Antimony pieces, 5mm (0.2in) & down, 99.9999% (metals basis), Thermo Scientific Chemicals
Catalog number011676.09
Price (USD)/ Each
157.00
-
Add to cart
Chemical Identifiers
CAS7440-36-0
IUPAC Nameantimony
Molecular FormulaSb
InChI KeyWATWJIUSRGPENY-UHFFFAOYSA-N
SMILES[Sb]
View more
SpecificationsSpecification SheetSpecification Sheet
FormPieces
Total Metal Impurities≤0.0001% (1 ppm)
Appearance (Color)Silver to grey
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only