Palladium, 5% on barium carbonate powder, reduced, Thermo Scientific Chemicals
Palladium, 5% on barium carbonate powder, reduced, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Palladium, 5% on barium carbonate powder, reduced, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Catalog number 011721.06
also known as 011721-06
Price (USD)/ Each
-
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPalladium
ColorGray
Name Note5% on barium carbonate powder
CAS7440-05-3
Health Hazard 1H302
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only