Palladium, 5% on barium sulfate powder, reduced, Thermo Scientific Chemicals
Palladium, 5% on barium sulfate powder, reduced, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Palladium, 5% on barium sulfate powder, reduced, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
2.5 g
25 g
Catalog number 011722.22
also known as 011722-22
Price (USD)/ Each
-
Quantity:
100 g
Request bulk or custom format
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialPalladium
ColorGray to Black
Name Note5% on barium sulfate powder
CAS7440-05-3
Health Hazard 1H302+H332
View more

Palladium, 5% on barium sulfate powder reduced usually supported on Barium sulfate, or an appropriate form of carbon, and useful catalyst for hydrogenations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only