Ruthenium, 5% on activated carbon powder, reduced, Thermo Scientific Chemicals
Ruthenium, 5% on activated carbon powder, reduced, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ruthenium, 5% on activated carbon powder, reduced, Thermo Scientific Chemicals

Aromatic ring hydrogenation
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number 011748.22
also known as 011748-22
Price (USD)/ Each
541.65
Online exclusive
602.00 
Save 60.35 (10%)
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
541.65
Online exclusive
602.00 
Save 60.35 (10%)
Add to cart
Ruthenium, 5% on activated carbon powder, reduced, Thermo Scientific Chemicals
Catalog number011748.22
Price (USD)/ Each
541.65
Online exclusive
602.00 
Save 60.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7440-18-8
IUPAC Nameruthenium
Molecular FormulaRu
InChI KeyKJTLSVCANCCWHF-UHFFFAOYSA-N
SMILES[Ru]
View more
SpecificationsSpecification SheetSpecification Sheet
Elemental AnalysisRu: 4.5 to 5.5%
Appearance (Color)Black
Loss on DryingWater: 5%, max
FormPowder
It is used in hydrogenation of aromatic ring systems, particularly the hindered systems. It finds wide use for the reduction of aliphatic carbonyl compounds and carboxylic acids to alcohols, hydrogenation of anilines to cyclohexylamines, synthesis of ammonia, depolymerization of carbohydrates, Fischer-Tropsch synthesis, and hydrogenation of glucose to produce sorbitol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only