Rhodium, 1% on alumina powder, reduced, Thermo Scientific Chemicals
Rhodium, 1% on alumina powder, reduced, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Rhodium, 1% on alumina powder, reduced, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number 011769.18
also known as 011769-18
Price (USD)/ Each
-
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialRhodium
Name Note1% on alumina powder, Reduced
CAS7440-16-6
Health Hazard 1H228
Health Hazard 2GHS H Statement
H335
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only