Thermo Scientific Chemicals

Ruthenium(IV) oxide, anhydrous, Premion™, 99.95% (metals basis), Ru 75.2% min, Thermo Scientific Chemicals

Catalog number: 011804.02
0.5 g, Each
Thermo Scientific Chemicals

Ruthenium(IV) oxide, anhydrous, Premion™, 99.95% (metals basis), Ru 75.2% min, Thermo Scientific Chemicals

Catalog number: 011804.02
0.5 g, Each
Quantity
Catalog number: 011804.02
also known as 011804-02
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12036-10-1
IUPAC Name
dioxoruthenium
Molecular Formula
O2Ru
InChI Key
WOCIAKWEIIZHES-UHFFFAOYSA-N
SMILES
O=[Ru]=O
Total Metal Impurities
0.05% max.
Assay (unspecified)
Ru 75.2% min

Description

The most often used oxygen catalyst is ruthenium(IV) oxide. It is applied as an electrochemical super capacitor material, and it has high capacity to store charge. RuO2 is used as catalyst in the Deacon process which produces chlorine through the oxidation of hydrogen chloride. Owing to the high charge transfer capability, it is used as active material in super capacitors. The dispersion property of RuO2 is used to increase the capacitance of nanocomposite electrode materials for supercapacitors. RuO2 is extensively used for the coating of titanium anodes, and as a catalyst and doping agent, for use in the removal of hydrogen sulfide from oil refineries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text