Thermo Scientific Chemicals

Ruthenium(III) chloride oxide, ammoniated, Thermo Scientific Chemicals

Catalog number: 011806.06
5 g, Each
Thermo Scientific Chemicals

Ruthenium(III) chloride oxide, ammoniated, Thermo Scientific Chemicals

Catalog number: 011806.06
5 g, Each
Quantity
Catalog number: 011806.06
also known as 011806-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
11103-72-3
IUPAC Name
tris(λ²-ruthenium(2+)) tetradecaamine dihydrate hexachloride
Molecular Formula
Cl6H46N14O2Ru3
InChI Key
JQJSTVUROJELSR-UHFFFAOYSA-H
SMILES
N.N.N.N.N.N.N.N.N.N.N.N.N.N.O.O.[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Ru++].[Ru++].[Ru++]
Total Metal Impurities
≤0.1%
Appearance (Color)
Red to red-brown
Form
Powder or crystals or crystalline powder
Assay from Supplier's CofA
≥34.8% Ruthenium

Description

Microscopic stainRuthenium(III) chloride oxide, ammoniated is used as a reagent on plant material for staining pectin, gum, animal tissues and bacteria. It is also used in histology to stain aldehyde fixed mucopolysaccharides. Further, it serves as a pharmacological tool to study specific cellular mechanisms. It serves as an inhibitor of mitochondrial calcium uniporter. In addition, it is involved in reducing airway inflammation arises by pepper spray.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text