Sodium hexachlororhodate(III) dodecahydrate, Rh 17.1%, Thermo Scientific Chemicals
Sodium hexachlororhodate(III) dodecahydrate, Rh 17.1%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Sodium hexachlororhodate(III) dodecahydrate, Rh 17.1%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number 011809.03
also known as 011809-03
Price (USD)/ Each
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS14972-70-4
IUPAC Nametrisodium hexachlororhodiumheptakis(ylium) dodecahydrate
Molecular FormulaCl6H24Na3O12Rh
InChI KeyJCMYDKHYUGZPJG-UHFFFAOYSA-H
SMILESO.O.O.O.O.O.O.O.O.O.O.O.[Na+].[Na+].[Na+].Cl[Rh+7](Cl)(Cl)(Cl)(Cl)Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)Rh 17.1% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only