Thermo Scientific Chemicals

Ammonium hexachlororhodate(III) hydrate, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 011812.06
5 g, Each
Thermo Scientific Chemicals

Ammonium hexachlororhodate(III) hydrate, Premion™, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog number: 011812.06
5 g, Each
Quantity
Catalog number: 011812.06
also known as 011812-06
Price (USD)
Have Questions?

Chemical Identifiers

CAS
15336-18-2
IUPAC Name
triammonium hexachlororhodiumheptakis(ylium)
Molecular Formula
Cl6H12N3Rh
InChI Key
QNMMDVLCBKRUJU-UHFFFAOYSA-K
SMILES
[NH4+].[NH4+].[NH4+].Cl[Rh+7](Cl)(Cl)(Cl)(Cl)Cl
Total Metal Impurities
0.01% max.

Description

Ammonium hexachlororhodate(III) hydrate is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text