Thermo Scientific Chemicals

Rhodium(III) oxide, anhydrous, 99.9% (metals basis), Rh 80.6% min, Thermo Scientific Chemicals

Catalog number: 011814.02
0.5 g, Each
Thermo Scientific Chemicals

Rhodium(III) oxide, anhydrous, 99.9% (metals basis), Rh 80.6% min, Thermo Scientific Chemicals

Catalog number: 011814.02
0.5 g, Each
Quantity
Catalog number: 011814.02
also known as 011814-02
Price (USD)
Have Questions?

Chemical Identifiers

CAS
12036-35-0
IUPAC Name
dirhodium(3+) trioxidandiide
Molecular Formula
O3Rh2
InChI Key
XBBXDTCPEWHXKL-UHFFFAOYSA-N
SMILES
[O--].[O--].[O--].[Rh+3].[Rh+3]
Total Metal Impurities
≤0.1%
Appearance (Color)
Black
Assay (special)
Rh 80.6-81.0%
Form
Powder

Description

Use in the catalytic converters of automobiles. It is also used as an industrial catalyst in the manufacturing of chemical intermediates such as oxo-alcohols, as well as nitric acid and ethanoic (acetic) acid production. It is also an area of interest for new capacitor electrode material researchers, dynamic random access memory (DRAMs) and non volatile random access memories (FeRAMs) makers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text