Rhodium(III) chloride, anhydrous, 99.9% (metals basis), Rh 48.7% min, Thermo Scientific Chemicals
Rhodium(III) chloride, anhydrous, 99.9% (metals basis), Rh 48.7% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Rhodium(III) chloride, anhydrous, 99.9% (metals basis), Rh 48.7% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
0.25 g
1 g
5 g
Catalog number 011815.01
also known as 011815-01
Price (USD)/ Each
-
Quantity:
0.25 g
Request bulk or custom format
Chemical Identifiers
CAS10049-07-7
IUPAC Namerhodium(3+) trichloride
Molecular FormulaCl3Rh
InChI KeySONJTKJMTWTJCT-UHFFFAOYSA-K
SMILES[Cl-].[Cl-].[Cl-].[Rh+3]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
It is used in homogeneous catalysis, for example in the industrial prodcution of acetic acid. It is used to coat optic fibres and optical mirrors, and for crucibles, thermocouple elements and headlight reflectors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only