Thermo Scientific Chemicals

Rhodium(III) bromide hydrate, Thermo Scientific Chemicals

Catalog number: 011816.03
1 g, Each
Thermo Scientific Chemicals

Rhodium(III) bromide hydrate, Thermo Scientific Chemicals

Catalog number: 011816.03
1 g, Each
Quantity
Catalog number: 011816.03
also known as 011816-03
Price (USD)
Have Questions?

Chemical Identifiers

CAS
123333-87-9
IUPAC Name
rhodium(3+) tribromide
Molecular Formula
Br3Rh
InChI Key
MMRXYMKDBFSWJR-UHFFFAOYSA-K
SMILES
[Br-].[Br-].[Br-].[Rh+3]
Assay (unspecified)
Rh 23.6% min

Description

Rhodium(III) bromide hydrate is used to prepare coordination complex, trans-dibromotetrakis(pyridine)rhodium(III) bromide hexahydrate. It is also involved in the preparation of rhodium nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text