Tellurium(IV) oxide, 99.995% (metals basis), Thermo Scientific Chemicals
Tellurium(IV) oxide, 99.995% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Tellurium(IV) oxide, 99.995% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Catalog number 011821.30
also known as 011821-30
Price (USD)/ Each
798.00
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
798.00
Add to cart
Tellurium(IV) oxide, 99.995% (metals basis), Thermo Scientific Chemicals
Catalog number011821.30
Price (USD)/ Each
798.00
-
Add to cart
Chemical Identifiers
CAS7446-07-3
IUPAC Name(oxo-λ⁴-tellanylidene)oxidane
Molecular FormulaO2Te
InChI KeyLAJZODKXOMJMPK-UHFFFAOYSA-N
SMILESO=[Te]=O
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.005% max.
Tellurium(IV) oxide is also used in devices which can convert light into sound (acousto-optic material). Glasses made of tellurium oxide have high refractive indices and transmit into mid-IR region. It is used to make glasses with special properties. It is useful in optical waveguides and optical fiber amplification.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only