Thermo Scientific Chemicals

Tungsten(VI) oxide, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog number: 011828.A3
2 kg, Each
Thermo Scientific Chemicals

Tungsten(VI) oxide, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog number: 011828.A3
2 kg, Each
Quantity
Catalog number: 011828.A3
also known as 011828-A3
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1314-35-8
IUPAC Name
trioxotungsten
Molecular Formula
O3W
InChI Key
ZNOKGRXACCSDPY-UHFFFAOYSA-N
SMILES
O=[W](=O)=O
Form
Powder
Appearance (Color)
Yellow
Total Metal Impurities
≤0.2% (2000 ppm)
Particle Size
10-25 Micron average

Description

It is used in tungsten and tungstate manufacturing which are used as X-ray screens and for fire proofing fabrics. It is used as a ceramic pigment. Nanowires of Tungsten (VI) oxide are capable of absorbing a higher percentage of the sun radiation since it absorbs blue light.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text