Thermo Scientific Chemicals

Tungsten sulfide, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog number: 011829.18
50 g, Each
Thermo Scientific Chemicals

Tungsten sulfide, 99.8% (metals basis), Thermo Scientific Chemicals

Catalog number: 011829.18
50 g, Each
Quantity
Catalog number: 011829.18
also known as 011829-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12138-09-9
IUPAC Name
disulfanylidenetungsten
Molecular Formula
S2W
InChI Key
ITRNXVSDJBHYNJ-UHFFFAOYSA-N
SMILES
S=[W]=S
Total Metal Impurities
0.2% max.

Description

Tungsten sulfide is used as a component of catalysts for hydrotreating of crude oil, hydrodesulfurization and hydrodenitrification reactions. It finds application as solid-state secondary lithium battery cathodes and other electrochemical devices. It is found that it forms inorganic nanotubes, which is better reinforcing agents than carbon nanotubes. Tungsten sulfide addition to epoxy resin improves adhesion, fracture toughness and strain energy release rate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text