Thermo Scientific Chemicals

Ammonium molybdate (para) tetrahydrate, ACS, 81-83% as MoO{3}, Thermo Scientific Chemicals

Catalog number: 011831.18
50 g, Each
Thermo Scientific Chemicals

Ammonium molybdate (para) tetrahydrate, ACS, 81-83% as MoO{3}, Thermo Scientific Chemicals

Catalog number: 011831.18
50 g, Each
Quantity
Catalog number: 011831.18
also known as 011831-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12054-85-2
IUPAC Name
ammonium hydrate molybdenum oxidandiide
Molecular Formula
H6MoNO2
InChI Key
XCOMAVCHADVANQ-UHFFFAOYSA-O
SMILES
[NH4+].O.[O--].[Mo]
Heavy Metals
(OES-ICP) 0.001%
Impurity content
Arsenate, phosphate, and silicate (as SiO₂) 0.001%
Impurity content
Chloride (as Cl) 0.002%
Impurity content
K 0.01%
Impurity content
Mg 0.005%

Description

Ammonium molybdate (para) tetrahydrate, is used in photography,analytical lab reagent for phosphate,lead,arsenate determination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text