Ammonium molybdate (para) tetrahydrate, ACS, 81-83% as MoO{3}, Thermo Scientific Chemicals
Ammonium molybdate (para) tetrahydrate, ACS, 81-83% as MoO{3}, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ammonium molybdate (para) tetrahydrate, ACS, 81-83% as MoO{3}, Thermo Scientific Chemicals

Ammonium molybdate (para) tetrahydrate, H24Mo7N6O24.4H2O, CAS Number-12054-85-2, Ammonium paramolybdate, 2kg, Powder, 81-83%, 2.498, 234-722-4, 1235.86 (1163.79 Anhydrous), ACS, H302-H315-H319-H335, P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
50 g
100 g
2 kg
Catalog number 011831.36
also known as 011831-36
Price (USD)/ Each
168.65
Online exclusive
187.00 
Save 18.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
168.65
Online exclusive
187.00 
Save 18.35 (10%)
Add to cart
Ammonium molybdate (para) tetrahydrate, ACS, 81-83% as MoO{3}, Thermo Scientific Chemicals
Catalog number011831.36
Price (USD)/ Each
168.65
Online exclusive
187.00 
Save 18.35 (10%)
-
Add to cart
Chemical Identifiers
CAS12054-85-2
IUPAC Nameammonium hydrate molybdenum oxidandiide
Molecular FormulaH6MoNO2
InChI KeyXCOMAVCHADVANQ-UHFFFAOYSA-O
SMILES[NH4+].O.[O--].[Mo]
View more
SpecificationsSpecification SheetSpecification Sheet
Heavy Metals(OES-ICP) 0.001%
Impurity contentArsenate, phosphate, and silicate (as SiO₂) 0.001%
Impurity contentChloride (as Cl) 0.002%
Impurity contentK 0.01%
Impurity contentMg 0.005%
View more
Ammonium molybdate (para) tetrahydrate, is used in photography,analytical lab reagent for phosphate,lead,arsenate determination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only