Molybdenum(V) chloride, 99.6% (metals basis), Thermo Scientific Chemicals
Molybdenum(V) chloride, 99.6% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals

Molybdenum(V) chloride, 99.6% (metals basis), Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
50 g
250 g
1 kg
Catalog number 011832.06
also known as 011832-06
Price (USD)/ Each
39.65
Online exclusive
44.30 
Save 4.65 (11%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
39.65
Online exclusive
44.30 
Save 4.65 (11%)
Add to cart
Molybdenum(V) chloride, 99.6% (metals basis), Thermo Scientific Chemicals
Catalog number011832.06
Price (USD)/ Each
39.65
Online exclusive
44.30 
Save 4.65 (11%)
-
Add to cart
Chemical Identifiers
CAS10241-05-1
IUPAC Namemolybdenum(5+) pentachloride
Molecular FormulaCl5Mo
InChI KeyGICWIDZXWJGTCI-UHFFFAOYSA-I
SMILES[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Mo+5]
View more
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.4% max.
Molybdenum pentachloride is employed as a brazing and soldering flux for monel, tungsten alloys and copper. It has been proven to be an efficient catalyst for the chlorination of aromatic rings and oxidative coupling reactions. It is a strong oxidizing agent. In the Friedel-Craft reaction, molybdenum pentachloride is used as a good lewis acid. Molybdenum pentachloride is used to prepare molybdenum by vapor deposition method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only