Thermo Scientific Chemicals

Molybdenum(V) chloride, 99.6% (metals basis), Thermo Scientific Chemicals

Catalog number: 011832.18
50 g, Each
Thermo Scientific Chemicals

Molybdenum(V) chloride, 99.6% (metals basis), Thermo Scientific Chemicals

Catalog number: 011832.18
50 g, Each
Quantity
Catalog number: 011832.18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
10241-05-1
IUPAC Name
molybdenum(5+) pentachloride
Molecular Formula
Cl5Mo
InChI Key
GICWIDZXWJGTCI-UHFFFAOYSA-I
SMILES
[Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Mo+5]
Total Metal Impurities
0.4% max.

Description

Molybdenum pentachloride is employed as a brazing and soldering flux for monel, tungsten alloys and copper. It has been proven to be an efficient catalyst for the chlorination of aromatic rings and oxidative coupling reactions. It is a strong oxidizing agent. In the Friedel-Craft reaction, molybdenum pentachloride is used as a good lewis acid. Molybdenum pentachloride is used to prepare molybdenum by vapor deposition method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text